MilliporeSigma
  • Treatment of a BTo-X-contaminated gas stream with a biotrickling filter inoculated with microbes bound to a wheat bran/red wood powder/diatomaceous earth carrier.
Treatment of a BTo-X-contaminated gas stream with a biotrickling filter inoculated with microbes bound to a wheat bran/red wood powder/diatomaceous earth carrier.
Bioresource technology (2010-06-26)
Jian M Chen, Run Y Zhu, Wei B Yang, Li L Zhang
ABSTRACT
Microbes bound to a wheat bran/red wood powder/diatomaceous earth carrier were used as inoculants for a biotrickling filter (BTF) for treating gases contaminated with a mixture of benzene, toluene, and o-xylene (BTo-X). An overall removal efficiency of more than 87.9% was achieved after a start-up period of as low as 4days. At BTo-X loading rates (LRs) below 60.0g/m(3)h, the BTF's performance was similar for EBRTs of 90, 60, 45 and 30s with an elimination capacity (EC) almost approaching the LR; stable REs above 91.3% for benzene and toluene and above 82.8% for o-xylene were achieved. A maximum EC of 97.7g/m(3)h was obtained at inlet load of 146.4g/m(3)h. The mass ratio of carbon dioxide produced to the BTo-X removed was approximately 2.62, which confirmed complete degradation of BTo-X. The results demonstrate that microbes bound to a solid carrier can be an alternative to traditional liquid inoculums applied in BTFs and highlight their potential applicability to BTF technologies.
Product Description
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
Xylenes, reagent grade
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Sigma-Aldrich
Xylenes, SAJ special grade, ≥80.0%
Sigma-Aldrich
Xylenes, SAJ first grade, ≥80.0%
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Xylenes, histological grade
Sigma-Aldrich
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
o-Xylene, anhydrous, 97%