Merck
  • Analysis of phenprocoumon and its hydroxylated and conjugated metabolites in human urine by high-performance liquid chromatography after solid-phase extraction.
Analysis of phenprocoumon and its hydroxylated and conjugated metabolites in human urine by high-performance liquid chromatography after solid-phase extraction.
Journal of chromatography (1990-09-14)
P M Edelbroek, G M van Kempen, T J Hessing, F A de Wolff
ABSTRACT
The anticoagulant phenprocoumon is mainly metabolized in humans to hydroxylated metabolites and their glucuronides. A method is described for the determination of phenprocoumon, 4'-hydroxyphenprocoumon, 6-hydroxyphenprocoumon, 7-hydroxyphenprocoumon, and their glucuronide and sulphate conjugates in human urine. Reversed-phase high-performance liquid chromatography is performed after selective extraction with disposable quaternary amine columns of untreated, and beta-glucuronidase- or sulphatase-treated urine samples. Urinary excretion data are presented for total, glucuronidated, sulphated and free phenprocoumon, 4'-hydroxyphenprocoumon, 6-hydroxyphenprocoumon and 7-hydroxyphenprocoumon in twelve patients after an average daily dosage of 1.3-4.2 mg phenprocoumon.
Sigma-Aldrich
4-Methylumbelliferyl sulfate potassium salt, sulfatase substrate