MilliporeSigma
  • Multicomponent reaction of imidazo[1,5-a]pyridine carbenes with aldehydes and dimethyl acetylenedicarboxylate or allenoates: a straightforward approach to fully substituted furans.
Multicomponent reaction of imidazo[1,5-a]pyridine carbenes with aldehydes and dimethyl acetylenedicarboxylate or allenoates: a straightforward approach to fully substituted furans.
The Journal of organic chemistry (2010-09-14)
Huan-Rui Pan, Yong-Jia Li, Cai-Xia Yan, Juan Xing, Ying Cheng
ABSTRACT
The facile three-component reactions of N,N-substituted imidazo[1,5-a]pyridine carbenes, namely imidazo[1,5-a]pyridin-3-ylidenes, with aldehydes and DMAD or allenoates were disclosed. Both reactions proceeded via tandem nucleophilic addition, [3 + 2]-cycloaddition, and ring transformation to produce different 4-[(2-pyridyl)methyl]aminofuran derivatives generally in moderate yields. This work not only provided the first example of the application of imidazo[1,5-a]pyridin-3-ylidenes in organic synthesis but also developed a straightforward approach to fully substituted furans that are not easily accessible by other methods.
Sigma-Aldrich
Dimethyl acetylenedicarboxylate, 95%