MilliporeSigma
The metabolism of olaquindox in rats, chickens and pigs.
Toxicology letters (2010-10-27)
Zhao-Ying Liu, Ling-Li Huang, Xiao-Ni Zhou, Dong-Mei Chen, Yan-Fei Tao, Hua-Hai Zhang, Zong-Hui Yuan
ABSTRACT
Olaquindox is a growth-promoting feed additive for food-producing animals. Its toxicities were reported to be closely related to the metabolism. To provide the interpretation of toxicities in animals, this study explored the metabolism of olaquindox in rats, chickens and pigs of different genders by qualitative metabolite profiling. Animals were fed olaquindox in an oral dose, and then their urine, plasma, feces, liver, kidney and muscle were collected. Liquid chromatography combined with hybrid ion trap/time-of-flight mass spectrometry was used for structural investigation and identification of metabolites. The structures of metabolites were elucidated based on the accurate MS² spectra and comparison of their changes in accurate molecular masses and fragment ions with those of parent drug or metabolite. A total of 18, 18 and 16 metabolites of rats, chickens and pigs were identified, respectively. Among the identified metabolites, 8 known metabolites were confirmed as an early study had stated, and 15 metabolites were found for the first time in vivo. The major metabolic pathways of olaquindox were proposed to be N-O reduction and oxidation of hydroxyl to carboxylic acid followed by N-O reduction. The qualitative species difference on the metabolite profiles of olaquindox among the three species was observed. However, metabolite profiles of olaquindox appeared to be qualitatively similar between female and male for the same species. The proposed metabolic pathways of olaquindox in animals will provide comprehensive data to clarify the metabolism of olaquindox among different species, and will give scientific explanation for toxicities and residues of olaquindox.
Product Description
Supelco
Olaquindox, VETRANAL®, analytical standard