MilliporeSigma
  • Preparation and properties of a coated slow-release and water-retention biuret phosphoramide fertilizer with superabsorbent.
Preparation and properties of a coated slow-release and water-retention biuret phosphoramide fertilizer with superabsorbent.
Journal of agricultural and food chemistry (2010-12-16)
Shuping Jin, Guoren Yue, Lei Feng, Yuqi Han, Xinghai Yu, Zenghu Zhang
ABSTRACT
In this investigation, a novel water-insoluble slow-release fertilizer, biuret polyphosphoramide (BPAM), was formulated and synthesized from urea, phosphoric acid (H(3)PO(4)), and ferric oxide (Fe(2)O(3)). The structure of BPAM was characterized by Fourier transform infrared (FTIR) spectroscopy. Subsequently, a coated slow-release BPAM fertilizer with superabsorbent was prepared by ionic cross-linked carboxymethylchitosan (the core), acrylic acid, acrylamide, and active carbon (the coating). The variable influences on the water absorbency were investigated and optimized. Component analysis results showed that the coated slow-release BPAM contained 5.66% nitrogen and 11.7% phosphorus. The property of water retention, the behavior of slow release of phosphorus, and the capacity of adsorption of cations were evaluated, and the results revealed that the product not only had good slow-release property and excellent water retention capacity but also higher adsorption capacities of cations in saline soil.
Sigma-Aldrich
Biuret, 98% (Contains ≤10%(w/w) Triuret)