MilliporeSigma
  • Localization of lipoxygenase activity on the oil bodies and in protoplasts using a novel fluorescence imaging method.
Localization of lipoxygenase activity on the oil bodies and in protoplasts using a novel fluorescence imaging method.
Plant physiology and biochemistry : PPB (2010-12-21)
Mukesh K Yadav, Satish C Bhatla
ABSTRACT
Lipoxygenase (linoleate:oxygen oxidoreductase; EC 1.13.11.12; LOX) catalyzes oxygenation of polyenoic fatty acids, which precedes the degradation of storage lipids during seed germination in sunflower. In the present work, it has been confirmed that 2',7'-dichlorodihydrofluorescein diacetate (H(2)DCFDA) produces fluorescence in presence of lipid hydroperoxides (LOX reaction products). This work provides new information on spatial localization of transiently enhanced LOX activity in protoplasts from 5 d old seedling cotyledons of sunflower (Helianthus annuus L. cv. Morden) by exploiting H(2)DCFDA as a probe for fluorescence detection from LOX activity sites. Use of LOX inhibitors [nordihydroguaiaretic acid (NDGA) and propyl gallate (PG)] confirms oil bodies as LOX activity sites. Oil body surface has been shown to possess LOX activity in 5 d old seedling cotyledons.
Sigma-Aldrich
Propyl gallate, for microscopy, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, powder
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Supelco
Propyl gallate, Pharmaceutical Secondary Standard; Certified Reference Material