MilliporeSigma
  • Biodegradation of fenvalerate and 3-phenoxybenzoic acid by a novel Stenotrophomonas sp. strain ZS-S-01 and its use in bioremediation of contaminated soils.
Biodegradation of fenvalerate and 3-phenoxybenzoic acid by a novel Stenotrophomonas sp. strain ZS-S-01 and its use in bioremediation of contaminated soils.
Applied microbiology and biotechnology (2010-12-25)
Shaohua Chen, Liu Yang, Meiying Hu, Jingjing Liu
ABSTRACT
A bacterial strain ZS-S-01, newly isolated from activated sludge, could effectively degrade fenvalerate and its hydrolysis product 3-phenoxybenzoic acid (3-PBA). Based on the morphology, physiological biochemical characteristics, and 16 S rDNA sequence, strain ZS-S-01 was identified as Stenotrophomonas sp. Strain ZS-S-01 could also degrade and utilize deltamethrin, beta-cypermethrin, beta-cyfluthrin, and cyhalothrin as substrates for growth. Strain ZS-S-01 was capable of degrading fenvalerate rapidly without a lag phase over a wide range of pH and temperature, even in the presence of other carbon sources, and metabolized it to yield 3-PBA, then completely degraded it. No persistent accumulative product was detected by HPLC and GC/MS analysis. Studies on biodegradation in various soils showed that strain ZS-S-01 demonstrated efficient degradation of fenvalerate and 3-PBA (both 50 mg·kg(-1)) with a rate constant of 0.1418-0.3073 d(-1), and half-lives ranged from 2.3 to 4.9 days. Compared with the controls, the half-lives for fenvalerate and 3-PBA reduced by 16.9-156.3 days. These results highlight strain ZS-S-01 may have potential for use in bioremediation of pyrethroid-contaminated environment.
Product Description
Supelco
β-Cyfluthrin, PESTANAL®, analytical standard
Supelco
Baythroid®, PESTANAL®, analytical standard