MilliporeSigma
Neuroendocrine contributions to sexual partner preference in birds.
Frontiers in neuroendocrinology (2011-02-01)
Elizabeth Adkins-Regan
ABSTRACT
A majority of birds are socially monogamous, providing exceptional opportunities to discover neuroendocrine mechanisms underlying preferences for opposite-sex partners where the sexes form extended affiliative relationships. Zebra finches have been the focus of the most systematic program of research to date in any socially monogamous animal. In this species, sexual partner preference can be partially or largely sex reversed with hormone manipulations during early development, suggesting a role for organizational hormone actions. This same conclusion emerges from research with Japanese quail, which do not form long-term pairs. In zebra finches, social experience manipulations during juvenile development also can sex reverse partner preference, either alone or in combination with an early hormone manipulation. Although there are several candidate brain regions where neural mechanisms could underlie these effects of hormones or social experience, the necessary research has not yet been done to determine their involvement. The neuroendocrinology of avian sexual partner preference is still frontier territory.
Sigma-Aldrich
Fadrozole hydrochloride, ≥98% (HPLC)