MilliporeSigma
  • Dioxygenases, key enzymes to determine the aglycon structures of fusicoccin and brassicicene, diterpene compounds produced by fungi.
Dioxygenases, key enzymes to determine the aglycon structures of fusicoccin and brassicicene, diterpene compounds produced by fungi.
Journal of the American Chemical Society (2011-02-09)
Yusuke Ono, Atsushi Minami, Motoyoshi Noike, Yusuke Higuchi, Tomonobu Toyomasu, Takeshi Sassa, Nobuo Kato, Tohru Dairi
ABSTRACT
Fusicoccin A and cotylenin A are structurally related diterpene glucosides and show a phytohormone-like activity. However, only cotylenin A induces the differentiation of human myeloid leukemia cells. Since the cotylenin A producer lost its ability to proliferate during preservation, a study on the relationship between structure and activity was carried out and a modified fusicoccin A with hydroxyl group at the 3-position showed a similar biological activity with that of cotylenin A. We then searched for an enzyme source that catalyzes the introduction of a hydroxyl group into the 3-position and found that brassicicene C, which is structurally related to fusicoccin A with hydroxyl group at the 3-position, was produced by Alternaria brassicicola ATCC96836. We recently cloned a brassicicene C biosynthetic gene cluster including the genes encoding fusicocca-2,10(14)-diene synthase and two cytochrome P450s, which were responsible for the formation of fusicocca-2,10(14)-diene-8β,16-diol. In this study, we report that a α-ketoglutarate dependent dioxygenase, the gene coding for which was located in the cluster, catalyzed a hydroxylation at the 3-position of fusicocca-2,10(14)-diene-8β,16-diol. On the other hand, a α-ketoglutarate-dependent dioxygenase, which had been identified in a fusicoccin A biosynthetic gene cluster, catalyzed the 16-oxidation of fusicocca-2,10(14)-diene-8β,16-diol to yield an aldehyde (8β-hydroxyfusicocca-1,10(14)-dien-16-al), although both dioxygenases had 51% amino acid sequence identity. These findings suggested that the dioxygenases played critical roles for the formation of the fusicoccin A-type and cotylenin A-/brassicicene C-type aglycons. Moreover, we showed that short-chain dehydrogenase/reductase located in the fusicoccin A biosynthetic gene cluster catalyzed the reduction of the aldehyde to yield fusicocca-1,10(14)-diene-8β,16-diol.
Sigma-Aldrich
Fusicoccin from Fusicoccum amygdali, ≥85% (HPLC)