MilliporeSigma
  • [Newly leaching method of copper from waste print circuit board using hydrochloric acid/n-butylamine/copper sulfate].
[Newly leaching method of copper from waste print circuit board using hydrochloric acid/n-butylamine/copper sulfate].
Huan jing ke xue= Huanjing kexue (2011-03-03)
Hong-Yan Wang, Zhao-Jie Cui, Ya-Wei Yao
ABSTRACT
A newly leaching method of copper from waste print circuit board was established by using hydrochloric acid-n-butylamine-copper sulfate mixed solution. The conditions of leaching were optimized by changing the hydrochloric acid, n-butylamine, copper sulfate,temperature and other conditions using copper as target mimics. The results indicated that copper could be leached completely after 8 h at 50 degrees C, hydrochloric acid concentration of 1.75 mol/L, n-butylamine concentration of 0.25 mol/L, and copper sulfate mass of 0.96 g. Under the conditions, copper leaching rates in waste print circuit board samples was up to 95.31% after 9 h. It has many advantages such as better effects, low cost, mild reaction conditions, leaching solution recycling.
Sigma-Aldrich
Butylamine, ≥99%
Sigma-Aldrich
Butylamine, 99.5%