MilliporeSigma
  • Acute intoxication caused by overdose of flunitrazepam and triazolam: high concentration of metabolites detected at autopsy examination.
Acute intoxication caused by overdose of flunitrazepam and triazolam: high concentration of metabolites detected at autopsy examination.
The American journal of forensic medicine and pathology (2011-03-05)
Akira Namera, Ryosuke Makita, Tatsuro Saruwatari, Aiko Hatano, Hiroaki Shiraishi, Masataka Nagao
ABSTRACT
A 52-year-old woman was found dead on the floor of the living room on the first floor of a house, which belonged to the man with whom she shared the house. On visiting the site, her clothes were found to be undisturbed. Packages of flunitrazepam (Silece, 2 mg/tablet) and triazolam (Halcion, 0.25 mg/tablet) were found strewn around the victim. Toxicological analysis was performed, and the concentrations of flunitrazepam, triazolam, and their metabolites in the victim's blood and urine were measured by high-performance liquid chromatography coupled with photodiode array and mass spectrometry. A high blood concentration of 7-aminoflunitrazepam was detected (1,270 ng/g), and further metabolites such as 7-acetamidoflunitrazepam, 7-acetamidodesmethylflunitrazepam, and 7-aminodesmethylflunitrazepam were detected in the blood and urine samples. In addition, 4-hydroxytriazolam and α-hydroxytriazolam were detected in her urine at a concentration of 950 and 12,100 ng/mL, respectively.On the basis of the autopsy findings and toxicology results of high concentrations of both flunitrazepam and triazolam derivatives, the cause of death was determined to be acute intoxication from flunitrazepam and triazolam.
Sigma-Aldrich
α-Hydroxytriazolam
Supelco
Triazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
7-Aminoflunitrazepam solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
7-Aminoflunitrazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
α-Hydroxytriazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®