MilliporeSigma
  • Development and validation of two methods based on high-performance liquid chromatography-tandem mass spectrometry for determining 1,2-benzopyrone, dihydrocoumarin, o-coumaric acid, syringaldehyde and kaurenoic acid in guaco extracts and pharmaceutical preparations.
Development and validation of two methods based on high-performance liquid chromatography-tandem mass spectrometry for determining 1,2-benzopyrone, dihydrocoumarin, o-coumaric acid, syringaldehyde and kaurenoic acid in guaco extracts and pharmaceutical preparations.
Journal of separation science (2011-03-05)
João C Gasparetto, Thais M Guimarães de Francisco, Francinete R Campos, Roberto Pontarolo
ABSTRACT
In this study, two HPLC-ESI-MS/MS methods were developed and validated for the determination of 1,2-benzopyrone (COU), o-coumaric acid (OCA), kaurenoic acid (KAU), syringaldehyde (SYR), and dihydrocoumarin (DIH) in guaco extracts and pharmaceutical preparations (syrup and oral solution). The chromatographic separation was achieved using a C18 XBridge 150×2.1-mm (5-μm particle size) column maintained at 25°C. The mobile phases consisted of a gradient of water and acetonitrile containing 0.05% formic acid or 5  mM ammonium formate for the positive and negative ion modes, respectively. All of the calibration curves showed excellent coefficients of correlation (r≥0.9970) over the ranges of 1.25-400  ng/mL for coumarin, 10-600  ng/mL for dihydrocoumarin, 5-250  ng/mL for KAU, and 25-500  ng/mL for o-coumaric acid and syringaldehyde. The range of recovery was 96.3-103% with an RSD% of <4.85% for intraday and interday precision. The results indicate that the developed methods are fast, efficient, and sensitive for the quantification of the guaco metabolites in extracts and pharmaceutical forms while avoiding purification and derivatization steps.
Sigma-Aldrich
Syringaldehyde, 98%
Sigma-Aldrich
Syringaldehyde, ≥98%, FG
Sigma-Aldrich
Kaurenoic acid, ≥95% (LC/MS-ELSD)