MilliporeSigma
Lithium-induced supramolecular hydrogel.
Chemical communications (Cambridge, England) (2011-03-17)
Jong S Park, Sooyeon Jeong, Dong Wook Chang, Jae Pil Kim, Ketack Kim, Eun-Kyoung Park, Ki-Won Song
ABSTRACT
We describe a novel anisotropic supramolecular gel made of cyclodextrin-dye, in which physical gelation is completed by lithium salt. Rheological experiment reveals the elastic behaviors of the hydrogel, and high ionic conductivity represents a good mobility of ions inside the gel matrix.
Sigma-Aldrich
γ-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, Wacker Chemie AG, ≥90.0% cyclodextrin basis (HPLC)