MilliporeSigma
  • α-Tocopheryl phosphate: uptake, hydrolysis, and antioxidant action in cultured cells and mouse.
α-Tocopheryl phosphate: uptake, hydrolysis, and antioxidant action in cultured cells and mouse.
Free radical biology & medicine (2011-03-30)
Keiko Nishio, Noriko Ishida, Yoshiro Saito, Yoko Ogawa-Akazawa, Mototada Shichiri, Yasukazu Yoshida, Yoshihisa Hagihara, Noriko Noguchi, John Chirico, Jeffrey Atkinson, Etsuo Niki
ABSTRACT
α-Tocopheryl phosphate (α-TP), a water-soluble analogue of α-tocopherol, is found in humans, animals, and plants. α-TP is resistant to both acid and alkaline hydrolysis and may exert its own function in this form in vivo. In this study, the uptake, hydrolysis, and antioxidant action of α-TP were measured using α-TP with a deuterated methyl group, CD(3), at position 5 of the chroman ring (α-TP(CD3)). The hydrolysis of α-TP(CD3) was followed by measuring α-tocopherol containing the CD(3) group, α-T(CD3), in comparison to unlabeled α-tocopherol, α-T(CH3). α-TP(CD3) was incubated with cultured cells, and the intracellular α-T(CD3) formed was measured with HPLC-ECD and GC-MS. α-TP(CD3) was also administered to mice for 4 weeks by mixing in the diet, and α-T(CD3) was measured in plasma, liver, brain, heart, and testis to compare with endogenous unlabeled α-T(CH3). It was found that α-TP(CD3) was taken in and hydrolyzed readily to α-T(CD3) in cultured cells and in mice. The hydrolysis of α-TP(CD3) in cell culture medium was not observed. α-TP protected primary cortical neuronal cells from glutamate-induced cytotoxicity, and α-TP given to mice reduced the levels of lipid peroxidation products in plasma and liver. These results suggest that α-TP is readily hydrolyzed in vivo to α-T, which acts as an antioxidant, and that α-TP may be used as a water-soluble α-T precursor in intravenous fluids, in eye drops, or as a dietary supplement.
Sigma-Aldrich
(±)-α-Tocopherol phosphate disodium salt, water-soluble α-tocopherol analog