MilliporeSigma
  • The need for enzymatic steering in abietic acid biosynthesis: gas-phase chemical dynamics simulations of carbocation rearrangements on a bifurcating potential energy surface.
The need for enzymatic steering in abietic acid biosynthesis: gas-phase chemical dynamics simulations of carbocation rearrangements on a bifurcating potential energy surface.
Journal of the American Chemical Society (2011-05-10)
Matthew R Siebert, Jiaxu Zhang, Srirangam V Addepalli, Dean J Tantillo, William L Hase
ABSTRACT
Abietic acid, a constituent of pine resin, is naturally derived from abietadiene --a process that requires four enzymes: one (abietadiene synthase) for conversion of the acyclic, achiral geranylgeranyl diphosphate to the polycyclic, chiral abietadiene (a complex process involving the copalyl diphosphate intermediate) and then three to oxidize a single methyl group of abietadiene to the corresponding carboxylic acid. In previous work (Nature Chem.2009, 1, 384), electronic structure calculations on carbocation rearrangements leading to abietadienyl cation revealed an interesting potential energy surface with a bifurcating reaction pathway (two transition-state structures connected directly with no intervening minimum), which links two products--one natural and one not yet isolated from Nature. Herein we describe direct dynamics simulations of the key step in the formation of abietadiene (in the gas phase and in the absence of the enzyme). The simulations reveal that abietadiene synthase must intervene in order to produce abietadiene selectively, in essence steering this reaction to avoid the generation of byproducts with different molecular architectures.
Sigma-Aldrich
Gum rosin, CP
Sigma-Aldrich
Gum rosin, natural resin
Sigma-Aldrich
Abietic acid, technical, ~75% (GC)