MilliporeSigma
Evaluation of soybean methanol fraction on acute inflammation.
Journal of medicinal food (2011-06-15)
Vanessa da Silva Carrara, Ciomar Bersani Amado, Juliana Oliveira de Melo, José Marcos Gontijo Mandarino, Diógenes Aparício Garcia Cortez
ABSTRACT
Soybeans have been of interest of researchers because of the presence of isoflavones, a subclass of flavonoids, which have demonstrated anti-inflammatory activity. The aim of this study was investigate the anti-inflammatory activity of the methanol fraction from soybean, which contains mainly isoflavone glucosides and malonylglucosides. The anti-inflammatory activity of the methanol fraction from soybean was studied using croton oil-induced mouse ear edema and carrageenan-induced pleurisy models. The methanol fraction inhibited the ear edema in a dose-dependent manner: 0.625 mg/kg by 44.23% (P<.05), 1.25 mg/kg by 60.68% (P<.01), and 2.5 mg/kg by 65.68% (P<.01). Myeloperoxidase enzyme activity was reduced at the dose of 2.5 mg/kg (64.79%, P<.05). No effects were seen on carrageenan-induced pleurisy at different doses of the methanol fraction (100 or 400 mg/kg). These results demonstrated that the methanol fraction containing conjugated isoflavones showed topical anti-inflammatory activity. There was no acute toxicity in Swiss mice after oral administration of the fraction, at doses of 1,000, 2,000, 3,000, and 4,000 mg/kg.
Sigma-Aldrich
Croton oil