MilliporeSigma
The antidiabetic effect of ginsenoside Rb2 via activation of AMPK.
Archives of pharmacal research (2011-08-04)
Kyoung-Tae Lee, Tae Woo Jung, Hak-Ju Lee, Seul-Gi Kim, Yu-Su Shin, Wan-Kyunn Whang
ABSTRACT
Ginsenosides, which are active compounds found in ginseng (Panax ginseng), are used as antidiabetic treatments. The aim of this study was to determine whether Rb2, a type of ginsenoside, regulates hepatic gluconeogenesis through AMP-activated protein kinase (AMPK) and the orphan nuclear receptor small heterodimer partner (SHP) in hyperlipidemic conditions used as an in vitro model of type 2 diabetes. Considering these results, we concluded that Rb2 may inhibit palmitate-induced gluconeogenesis via AMPK-induced SHP by relieving ER stress, a cause of gluconeogenesis.
Product Description
Supelco
Ginsenoside Rb2, analytical standard