MilliporeSigma
  • [Effects of eight environmental endocrine disruptors on insulin resistance in patients with polycystic ovary syndrome: a preliminary investigation].
[Effects of eight environmental endocrine disruptors on insulin resistance in patients with polycystic ovary syndrome: a preliminary investigation].
Nan fang yi ke da xue xue bao = Journal of Southern Medical University (2011-10-27)
Ting-ting Li, Liang-zhi Xu, Yong-heng Chen, Hong-mei Deng, Chun-ying Liang, Ying Liu, Xiao-fang Liu, Jing Zhang, De-ying Kang, Dong-sheng Qiu, Dai-wen Han, Rong Hu
ABSTRACT
To investigate the relationship between 8 endocrine-disrupting chemicals in the serum and insulin resistance in patients with polycystic ovary syndrome (PCOS). This study was conducted among 60 patients with PCOS, including 23 with insulin resistance (PCOS-IR) and 37 without insulin resistance (PCOS-NIR), and 29 non-PCOS women seeking medical attention for infertility or menstrual disorder (control group). The serum levels of 6 phthalic acid esters (PEAs), bisphenol A (BPA) and octylphenol (OP) were measured in all the subjects. The levels of PAEs, BPA and OP showed no significant differences between PCOS patients and the control group (P>0.05). The serum level of OP was significantly lower in patients PCOS-IR than in those with PCOS-NIR (47.89 ng/ml vs 60.24 ng/ml, P<0.05). PEAs and BPA do not produce obvious effect on the pathogenesis of PCOS or contribute to insulin resistance, but OP may play a role in insulin resistance in PCOS patients.
Sigma-Aldrich
Diethyl phthalate, SAJ special grade, ≥98.0%
Supelco
Diethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Diethyl phthalate, 99.5%
Sigma-Aldrich
Diethyl phthalate, ≥99%