MilliporeSigma
  • Repeated exposure of adult rats to Aroclor 1254 induces neuronal injury and impairs the neurochemical manifestations of the NMDA receptor-mediated intracellular signaling in the hippocampus.
Repeated exposure of adult rats to Aroclor 1254 induces neuronal injury and impairs the neurochemical manifestations of the NMDA receptor-mediated intracellular signaling in the hippocampus.
Neurotoxicology (2011-11-03)
Wojciech Hilgier, Jerzy W Łazarewicz, Lidia Strużynska, Małgorzata Frontczak-Baniewicz, Jan Albrecht
ABSTRACT
Aroclor 1254 is a mixture of polychlorinated biphenyls (PCBs), a class of environmental toxins which cause a wide spectrum of neurotoxic effects. Learning and memory deficits are the profound effects of PCBs which may be related to hippocampal dysfunction. To get insight into the underlying neurochemical mechanisms, we employed the microdialysis technique to investigate the effect of repeated exposure of adult male Wistar rats to Aroclor 1254 (10mg/kg b.w., daily, ig., for 14days), on the neurochemical parameters of NMDA receptor-mediated glutamatergic signaling in the hippocampus in vivo assessed using the microdialysis technique. The results demonstrated that exposure to Aroclor 1254, which was associated with substantial neuronal damage and loss in the hippocampus, markedly decreased the NMDA-induced extracellular accumulation of newly loaded (45)CaCl(2), cGMP and glutamate, and reduced the basal content of the NO precursor, arginine, indicating inhibition of the NMDA/NO/cGMP pathway. Aroclor 1254 exposure also decreased the basal microdialysate content of glutamate and glutamine, which may cause inadequate supply of the neurotransmitter glutamate, while the level of two other neuroactive amino acids, aspartate or taurine was not affected by the exposure. The results underscore neuronal lesion and inhibition of NMDA receptor-mediated glutamatergic signaling in hippocampus as a potential major contributor to the cognitive deficits associated with exposure to PCB.
Product Description
Supelco
Aroclor 1254 solution, certified reference material, 50 mg/kg in transformer oil
Supelco
Aroclor 1254, ampule of 50 mg, analytical standard
Supelco
Aroclor 1254 solution, certified reference material, 500 mg/kg in transformer oil