MilliporeSigma
  • Melatonin membrane receptor type MT1 modulates cell-mediated immunity in the seasonally breeding tropical rodent Funambulus pennanti.
Melatonin membrane receptor type MT1 modulates cell-mediated immunity in the seasonally breeding tropical rodent Funambulus pennanti.
Neuroimmunomodulation (2011-11-10)
Raise Ahmad, Chandana Haldar, Sameer Gupta
ABSTRACT
Despite the evidence for melatonin membrane receptors (MT1R and MT2R) on lymphoid tissues in a wide range of seasonal breeders, their specific potency has never been compared and correlated with cell-mediated immunity. We used luzindole, a nonselective MT2R antagonist, and 4-phenyl-2-propionamidotetralin (4P-PDOT), a selective MT2R antagonist, to assess the potency of the melatonin receptors MT1 and MT2 in melatonin-induced immunity under both in vivo as well as in vitro conditions. Physiological doses (25 μg/100 g body weight in vivo and 100 and 500 pg/ml in vitro) of melatonin upregulated both MT1R and MT2R expression as well as splenocyte proliferation, while higher doses (100 and 500 μg/100 g body weight in vivo and 1 ng/ml in vitro) downregulated splenocyte proliferation and the expression of both receptors. Luzindole antagonized the expression of both MT1R and MT2R in a dose-dependent manner under in vivo as well as in vitro conditions, while 4P-PDOT blocked the expression of MT2R only during both experimental conditions. Splenocyte proliferation and IL-2 secretion (in vitro) followed the MT1R expression pattern, while the MT2R expression pattern showed no definite relation with either splenocyte proliferation or IL-2 secretion under in vivo and in vitro conditions. Immune function in tropical rodents is directly regulated by melatonin via its high-affinity membrane receptor MT1. MT1R plays a directive role in mediating splenocyte proliferation and IL-2 release, while the MT2R subtype appears not to be required for the immunoenhancing role of melatonin.
Sigma-Aldrich
Luzindole, ≥90%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00