MilliporeSigma
  • Deprotection of the S-trimethylacetamidomethyl (Tacm) group using silver tetrafluoroborate: application to the synthesis of porcine brain natriuretic peptide-32 (pBNP-32).
Deprotection of the S-trimethylacetamidomethyl (Tacm) group using silver tetrafluoroborate: application to the synthesis of porcine brain natriuretic peptide-32 (pBNP-32).
Chemical & pharmaceutical bulletin (1990-06-01)
M Yoshida, T Tatsumi, Y Fujiwara, S Iinuma, T Kimura, K Akaji, Y Kiso
ABSTRACT
Silver tetrafluoroborate (AgBF4) in trifluoroacetic acid (TFA) has been found to cleave the S-trimethyl-acetamidomethyl (Tacm) group or the S-acetamidomethyl (Acm) group without affecting other functional groups in a peptide chain. A newly isolated porcine brain natriuretic peptide-32 (pBNP-32) was synthesized by the combined use of the S-Tacm group and AgBF4 deprotection. The synthetic pBNP-32 was obtained in better yield by the AgBF4 procedure than by the standard I2 procedure. The synthetic pBNP-32 has the highest chick rectum relaxant activity among the known members of the atrial natriuretic peptide-brain natriuretic peptide (ANP-BNP) families. Somatostatin was also synthesized by the Fmoc-based solid-phase method using S-Tacm and AgBF4. In this synthesis, the recently developed reagent tetrafluoroboric acid (HBF4) was applied to cleave the peptide from the resin.
Sigma-Aldrich
Silver tetrafluoroborate, ≥99.99% trace metals basis
Sigma-Aldrich
Silver tetrafluoroborate, 98%