MilliporeSigma
  • Highly sensitive capillary electrophoresis analysis of N-linked oligosaccharides in glycoproteins following fluorescence derivatization with rhodamine 110 and laser-induced fluorescence detection.
Highly sensitive capillary electrophoresis analysis of N-linked oligosaccharides in glycoproteins following fluorescence derivatization with rhodamine 110 and laser-induced fluorescence detection.
Electrophoresis (2011-12-20)
Soichiro Ijiri, Kenichiro Todoroki, Hideyuki Yoshida, Takashi Yoshitake, Hitoshi Nohta, Masatoshi Yamaguchi
ABSTRACT
We describe a highly sensitive CE with laser-induced fluorescence (LIF) detection for the analysis of N-linked oligosaccharides in glycoproteins using rhodamine 110 as a fluorescence derivatization reagent. One CE separation is performed using a fused-silica capillary and neutral pH buffer conditions and allows for the separation of sialo-oligosaccharides according to the number of sialic acids. An alternate separation is performed using the same capillary and acidic pH buffer conditions, enabling the separation of asialo-oligosaccharides according to their sizes. The derivatization and separation conditions for the analysis of sialo- and asialo-oligosaccharides were optimized. Furthermore, we applied the proposed method for the analyses of N-linked sialo- and asialo-oligosaccharides in glycoproteins (ribonuclease B, fetuin, and recombinant human erythropoietin).
Sigma-Aldrich
Rhodamine 110 chloride, Dye content ≥75 %
Sigma-Aldrich
Rhodamine 110 chloride, suitable for fluorescence, BioReagent, ≥99.0% (UV)