MilliporeSigma
  • Pharmacological properties and related constituents of stem bark of Pterocarpus erinaceus Poir. (Fabaceae).
Pharmacological properties and related constituents of stem bark of Pterocarpus erinaceus Poir. (Fabaceae).
Asian Pacific journal of tropical medicine (2011-12-21)
Ouédraogo Noufou, Sawadogo Richard Wamtinga, Tibiri André, Bayet Christine, Lompo Marius, Hay A Emmanuelle, Koudou Jean, Dijoux Marie-Geneviève, Guissou Innocent Pierre
ABSTRACT
To screen methanol and dichloromethane extracts of stem bark of Pterocarpus erinaceus for anti-inflammatory, analgesic, in vitro antioxidant activities and phytochemical analysis. Anti-inflammatory activity was determined by using carrageenan induced-edema of mice paw and croton oil-induced edema of mice ear; analgesic effect was evaluated using acetic acid-induced writhing. Phytochemical screening of extracts was performed by thin layer chromatography. The chromatographic fractionation led to the isolation of main active components as friedelin, lupeol and epicathechin. The structures were established by TLC and nuclear magnetic resonance studies. Both methanol and dichloromethane extracts, friedelin, lupeol and epicatechin showed a significant anti-inflammatory effect using croton oil induced-ear edema. Furthermore, the action of dichloromethane extract was more important. At the doses of 100 and 200 mg/kg, the methanol extract was able to reduce the carrageenan induced-hind paw edema, while at the doses of 100, 200 and 400 mg/kg, it showed an important analgesic effect against writhing induced by acetic acid injection of 38.8%, 68.0% and 74.3%, respectively. Antioxidative properties of methanol extract and its dichloromethane and ethyl acetate fractions were assessed by using the 1,1-diphenyl-2-picrylhydrazyl method. The methanol extract showed the stronger radical scavenging activity than dichloromethane and ethyl acetate fractions, with an antiradical power of 5, 3.5 and 2 respectively. The main components isolated from these extracts as friedelin, lupeol and epicathechin were responsible of these activities. The results suggest that the stem bark extracts of Pterocarpus erinaceus possessed important anti-inflammatory, analgesic activities and strong antioxidant properties, therefore, they could be used as potential natural ingredients in the pharmaceutical industry.
Sigma-Aldrich
Friedelin, technical grade
Sigma-Aldrich
Croton oil
Sigma-Aldrich
Lupeol, ≥94%
Supelco
Lupeol, analytical standard