MilliporeSigma
  • Assessment of the working range and effect of sodium dichloroisocyanurate on Pseudomonas aeruginosa biofilms and planktonic cells.
Assessment of the working range and effect of sodium dichloroisocyanurate on Pseudomonas aeruginosa biofilms and planktonic cells.
Biofouling (2012-01-24)
Ari Morgenthau, Alexandru M Nicolae, Andrew E Laursen, Daniel A Foucher, Gideon M Wolfaardt, Martina Hausner
ABSTRACT
Sodium dichloroisocyanurate (NaDCC) is a chemical agent that acts against microorganisms in a manner similar to that of sodium hypochlorite by releasing free available chlorine. NaDCC has been approved by the WHO for the emergency treatment of water and by the US EPA for routine treatment of water. Previous studies assessing the effectiveness of NaDCC for the treatment of water implied that NaDCC should have a wide array of disinfecting effects beyond the treatment of planktonic cells in potable water. In this study the biocidal effects of NaDCC against Pseudomonas aeruginosa cells in different growth modes including planktonic cells and biofilms were explored. The data showed that a 60% dilution of the standard NaDCC solution was effective in the treatment of both P. aeruginosa planktonic cells and biofilms.
Sigma-Aldrich
Sodium dichloroisocyanurate, 96%
Sigma-Aldrich
Sodium dichloroisocyanurate dihydrate, 98.0% (AT)