MilliporeSigma
  • Protective effect of an elastase inhibitor in a neuromyelitis optica-like disease driven by a peptide of myelin oligodendroglial glycoprotein.
Protective effect of an elastase inhibitor in a neuromyelitis optica-like disease driven by a peptide of myelin oligodendroglial glycoprotein.
Multiple sclerosis (Houndmills, Basingstoke, England) (2012-02-22)
Katja Herges, Brigit A de Jong, Ilan Kolkowitz, Caitlin Dunn, Gil Mandelbaum, Rose M Ko, Alexander Maini, May H Han, Joep Killestein, Chris Polman, Alexandra L Goodyear, Jeffrey Dunn, Lawrence Steinman, Robert C Axtell
ABSTRACT
The pathology of neuromyelitis optica (NMO), in contrast to multiple sclerosis, comprises granulocyte infiltrates along extensive lengths of spinal cord, as well as optic nerve. Furthermore, IFN-β treatment worsens NMO. We recently found that experimental autoimmune encephalomyelitis (EAE) induced with Th17 cells is exacerbated by IFN-β, in contrast to disease induced with Th1 where treatment attenuated symptoms. This study demonstrates the similarities between NMO and Th17 EAE and how neutrophils mediate pathology in Th17 disease. Levels of blood biomarkers in NMO were assessed by Luminex and ELISA. Effects of IFN-β on neutrophils were assessed by culture assays and immunofluorescence. EAE was induced by transfer of myelin-specific Th1 or Th17 cells and treated with Sivelestat sodium hydrate, a neutrophil elastase inhibitor. We show Th17 cytokines, granulocyte chemokines, type 1 interferon and neutrophil elastase are elevated in patients with definitive NMO. In culture, we find that IFN-β stimulates neutrophils to release neutrophil elastase. In Th17 EAE, we demonstrate neutrophilic infiltration in the optic nerve and spinal cord which was not present in Th1 EAE. Blockade of neutrophil elastase with Sivelestat had efficacy in Th17 EAE but not Th1 EAE. The similarities between Th17 EAE and NMO indicate that this model represents several aspects of NMO. Neutrophils are critical in the pathologies of both Th17-EAE and NMO, and therefore blockade of neutrophil elastase is a promising target in treating NMO.
Sigma-Aldrich
Sivelestat sodium salt hydrate, ≥98% (HPLC), solid