MilliporeSigma
  • Visual observations and Raman spectroscopic studies of supercritical water oxidation of chlorobenzene in an anticorrosive fused-silica capillary reactor.
Visual observations and Raman spectroscopic studies of supercritical water oxidation of chlorobenzene in an anticorrosive fused-silica capillary reactor.
Environmental science & technology (2012-02-23)
Huicheng Liu, Zhiyan Pan
ABSTRACT
Supercritical water oxidation of chlorobenzene (CB) was studied using an anticorrosive fused-silica capillary reactor (FSCR) combined with a polarization microscope recorder system for visual observations and a Raman spectroscopic system for qualitative and quantitative analyses of the gaseous products. The effects of operating parameters, including the stoichiometric amount of oxidizer, temperature, and reaction time, on oxidation behavior were investigated. Our results show that a 100% conversion yield of CB and 100% CO(2) yield were achieved with a 150% stoichiometric amount of H(2)O(2) at 450 °C within 8 and 10 min, respectively. The conversion yield and the CO(2) yield both depend strongly on temperature, and the CO(2) yield is always less than the CB conversion yield under the same experimental conditions, suggesting that some carbon exists in intermediate products of incomplete oxidation, as confirmed by gas chromatography-mass spectrometry. Global kinetics analysis based on the complete conversion of CB to CO(2) showed that the reaction was first order. CB phase-changes in sub- and supercritical H(2)O-H(2)O(2) system in the FSCR were observed and recorded; CB eventually dissolved completely to form a homogeneous liquid solution above 326.1 °C. This method has great potential for use in the theoretical study of fluids and chemical reactions under elevated pressure-temperature conditions.
Sigma-Aldrich
Chlorobenzene, SAJ special grade, ≥99.5%
Sigma-Aldrich
Chlorobenzene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Chlorobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Chlorobenzene, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Supelco
Chlorobenzene, analytical standard
Sigma-Aldrich
Chlorobenzene, suitable for HPLC, 99.9%