Merck
  • Saquinavir inhibits the malaria parasite's chloroquine resistance transporter.
Saquinavir inhibits the malaria parasite's chloroquine resistance transporter.
Antimicrobial agents and chemotherapy (2012-02-23)
Rowena E Martin, Alice S Butterworth, Donald L Gardiner, Kiaran Kirk, James S McCarthy, Tina S Skinner-Adams
ABSTRACT
The antiretroviral protease inhibitors (APIs) ritonavir, saquinavir, and lopinavir, used to treat HIV infection, inhibit the growth of Plasmodium falciparum at clinically relevant concentrations. Moreover, it has been reported that these APIs potentiate the activity of chloroquine (CQ) against this parasite in vitro. The mechanism underlying this effect is not understood, but the degree of chemosensitization varies between the different APIs and, with the exception of ritonavir, appears to be dependent on the parasite exhibiting a CQ-resistant phenotype. Here we report a study of the role of the P. falciparum chloroquine resistance transporter (PfCRT) in the interaction between CQ and APIs, using transgenic parasites expressing different PfCRT alleles and using the Xenopus laevis oocyte system for the heterologous expression of PfCRT. Our data demonstrate that saquinavir behaves as a CQ resistance reverser and that this explains, at least in part, its ability to enhance the effects of CQ in CQ-resistant P. falciparum parasites.
Sigma-Aldrich
Saquinavir mesylate, ≥98% (HPLC), powder
Pricing and availability is not currently available.