- SnCl2/TiCl3-mediated deoximation of oximes in an aqueous solvent.
SnCl2/TiCl3-mediated deoximation of oximes in an aqueous solvent.
Molecules (Basel, Switzerland) (2012-03-03)
Mei-Huey Lin, Han-Jun Liu, Cheng-Yu Chang, Wei-Cheng Lin, Tsung-Hsun Chuang
PMID22382525
ABSTRACT
A simple procedure for SnCl(2)/TiCl(3)-mediated deoximation of ketoximes in an aqueous solvent is reported. Under the conditions developed in this effort, various ketones and aldehydes are produced in good to excellent yields.
Product Description
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tin(II) chloride dihydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tin standard solution, suitable for atomic absorption spectrometry, 1000 ppm Sn, 1 mg/mL Sn