MilliporeSigma
  • Analyzing the responses of saccharin in context with melatonin receptors on the melanophores of the fish Labeo rohita (Ham.).
Analyzing the responses of saccharin in context with melatonin receptors on the melanophores of the fish Labeo rohita (Ham.).
Journal of receptor and signal transduction research (2012-03-13)
M D Mubashshir, Fraz Ahmed, Safia Sumoona, Mohd Ovais
ABSTRACT
The hormone melatonin regulates the biological clock and assist in various other physiologies of vertebrates. Present work is intended to check the affinity of saccharin towards the melatonin receptors and the possible role of saccharin interference in the melatonin physiology. The present in vitro study is based on the working model of isolated scale melanophores in the dorso-lateral region of Labeo rohita. The pigment cells were incubated in the agonist and the antagonists within a limited time frame and subsequently their Melanophore Size Index (MSI) were calculated. The inferences were drafted through the observed signal transduction upshots in pigment translocations within the melanophores. Saccharin, in a wide dose range, has consistently induced a concentration-related aggregation similar to the aggregatory effect as shown by melatonin on the melanophores. Binding of saccharin with the receptors and eliciting its aggregatory effect is partially dependent on the release of neurotransmitters. The aggregatory effects were found to be significantly blocked by luzindole, K185, and prazosin, which are the potent melatonin receptor blockers, at the higher concentrations of saccharin. Hence, all the three subtypes of melatonin receptors viz. MT₁, MT₂, and MT₃ are participating in saccharin-mediated aggregations. Blocking by neomycin shows that Ca²⁺ ions are very crucial in dispensing the aggregatory effect of the sweetener. This research demands that an intensive and careful thorough study should be made about saccharin, specifically its effects upon melatonin physiology, before its unwarranted use as the food ingredients for human use.
Sigma-Aldrich
Luzindole, ≥90%