MilliporeSigma
  • Electronic spectra of gas-phase polycyclic aromatic nitrogen heterocycle cations: isoquinoline+ and quinoline+.
Electronic spectra of gas-phase polycyclic aromatic nitrogen heterocycle cations: isoquinoline+ and quinoline+.
The journal of physical chemistry. A (2012-04-14)
Viktoras Dryza, Julian A Sanelli, Evan G Robertson, Evan J Bieske
ABSTRACT
Electronic spectra of the gas-phase isoquinoline(+)-Ar and quinoline(+)-Ar complexes are recorded using photodissociation spectroscopy by monitoring the Ar loss channel. The D(3)←D(0) and D(4)←D(0) band origins for isoquinoline(+)-Ar are observed at 15245 ± 15 cm(-1) and 21960 ± 15 cm(-1), respectively, whereas for quinoline(+)-Ar they appear at 16050 ± 15 cm(-1) and 21955 ± 15 cm(-1), respectively. Strong vibronic progressions for the D(3)←D(0) band systems of both isoquinoline(+)-Ar and quinoline(+)-Ar are modeled and assigned in terms of ring deformation and carbon-carbon stretch vibrational modes using time-dependent density functional theory calculations in conjunction with Franck-Condon simulations. The properties of the isoquinoline(+) and quinoline(+) molecules are compared with those of the isoelectronic naphthalene(+) molecule. The existence of strong progressions in the visible spectra of isoquinoline(+)-Ar and quinoline(+)-Ar suggests that the corresponding isoquinoline(+) and quinoline(+) molecular cations are unlikely to be responsible for diffuse interstellar bands.
Sigma-Aldrich
Isoquinoline, 97%