MilliporeSigma
  • Acriflavine enhances radiosensitivity of colon cancer cells through endoplasmic reticulum stress-mediated apoptosis.
Acriflavine enhances radiosensitivity of colon cancer cells through endoplasmic reticulum stress-mediated apoptosis.
The international journal of biochemistry & cell biology (2012-05-09)
Min-Jin Lim, Ji-Yeon Ahn, Youngsoo Han, Chi-Ho Yu, Mi-Hyoung Kim, Sae-Lo-Oom Lee, Dae-Seog Lim, Jie-Young Song
ABSTRACT
Radiotherapy (RT) is one of the most effective tools in the clinical treatment of cancer. Because the tumor suppressor p53 plays a central role in radiation-mediated responses, including cell cycle-arrest and apoptosis, a number of studies have suggested that p53 could be a useful therapeutic target of anti-cancer agents. Accordingly, we sought to discover a new agent capable of increasing p53 activity. HCT116 colon cancer cells, containing wild-type p53, were stably transfected with a p53 responsive-luciferase (p53-Luc) reporter gene. A cell-based high-throughput screen of 7920 synthetic small molecules was performed in duplicate. Of the screened compounds, acriflavine (ACF) significantly increased p53-Luc activity in a concentration-dependent manner without causing toxicity. Pretreatment with ACF enhanced the induction of p53 protein expression and phosphorylation on serine 15 by γ-irradiation. Clonogenic assays showed that ACF pretreatment also potentiated radiation-induced cell death. The combination of irradiation and ACF treatment induced mitochondrial release of cytochrome c and significant activation of caspase-3 with PARP cleavage in colon cancer cells, demonstrating typical apoptotic cell death. Combined treatment with ACF and radiation increased the expression of Bax and Bad, while decreasing expression of Bcl-2. In addition, the ACF/radiation treatment combination induced endoplasmic reticulum (ER) stress responses mediated by IRE1α (inositol-requiring transmembrane kinase and endonuclease 1α), eIF-2α (eukaryotic initiation factor 2α), caspase-2/12, and CHOP (C/EBP homologous protein). The knockdown of IRE1α by siRNA inhibited the apoptotic cell death induced by ACF/radiation treatment. In vivo studies showed that combined treatment with ACF and radiation significantly inhibited the growth of tumors in colorectal cancer xenografted mice. These results indicate that ACF acts through p53-dependent mitochondrial pathways and ER stress signals, and could be a promising radiosensitizer.
Sigma-Aldrich
Acriflavine, fluorescent label
Sigma-Aldrich
Acriflavine, suitable for fluorescence, BioReagent, ≥90% (AT)
Sigma-Aldrich
Acriflavine hydrochloride, fluorescent dye