MilliporeSigma
  • Gene expression of ribosomal protein mRNA in Chironomus riparius: effects of endocrine disruptor chemicals and antibiotics.
Gene expression of ribosomal protein mRNA in Chironomus riparius: effects of endocrine disruptor chemicals and antibiotics.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2012-05-23)
Kiyun Park, Inn-Sil Kwak
ABSTRACT
Ribosomal protein genes are essential for cellular development. To examine the effects of ribosomal protein genes under various cellular stress conditions in chironomids, ribosomal protein S3 (RpS3) and S6 (RpS6) cDNA from Chironomus riparius were characterized and their expression was analyzed during development. A comparative and phylogenetic study among different orders of insects was carried out by analysis of sequence databases. C. riparius RpS3 was highly conserved at the protein level and shared over 85% amino acid identity with homologous sequences from other insects. RpS6 also showed approximately 80% amino acid identity. The RpS3 and S6 transcripts were present during different developmental stages but were most abundant during the embryonic stage. Furthermore, expression of the previously reported ribosomal proteins RpL11, L13, and L15, as well as RpS3 and S6 was analyzed following exposure to various concentrations of three endocrine disruptor chemicals (EDCs), di(2-ethylhexyl) phthalate, bisphenol A, and 4-nonylphenol (4NP), and the veterinary antibiotics (VAs) fenbendazole, sulfathiazole, and lincomycin. Only RpS3 gene expression was up-regulated significantly in response to EDCs and fenbendazole. However, the C. riparius ribosomal proteins showed a limited response to cellular stress, following exposure to EDCs and VAs.
Sigma-Aldrich
Lincomycin hydrochloride, 96.0-102.0% (HPLC)
Sigma-Aldrich
Fenbendazole, ≥98%
Sigma-Aldrich
Lincomycin hydrochloride, BioReagent, suitable for cell culture
Sigma-Aldrich
Lincomycin hydrochloride, ≥90% (TLC)
Supelco
Lincomycin hydrochloride monohydrate, VETRANAL®, analytical standard
Supelco
Fenbendazole, VETRANAL®, analytical standard
Supelco
Sulfathiazole
Supelco
Sulfathiazole, VETRANAL®, analytical standard
Supelco
4-Nonylphenol, PESTANAL®, analytical standard
Lincomycin hydrochloride, European Pharmacopoeia (EP) Reference Standard