MilliporeSigma
  • Phosphorylation of cyclic AMP-response element-binding protein (CREB) is influenced by melatonin treatment in pancreatic rat insulinoma β-cells (INS-1).
Phosphorylation of cyclic AMP-response element-binding protein (CREB) is influenced by melatonin treatment in pancreatic rat insulinoma β-cells (INS-1).
Journal of pineal research (2012-05-24)
Ivonne Bazwinsky-Wutschke, Sabine Wolgast, Eckhard Mühlbauer, Elke Albrecht, Elmar Peschke
ABSTRACT
The pineal hormone melatonin exerts its influence on the insulin secretion of pancreatic islets by a variety of signalling pathways. The purpose of the present study was to analyse the impact of melatonin on the phosphorylated transcription factor cAMP-response element-binding protein (pCREB). In pancreatic rat insulinoma β-cells (INS-1), pCREB immunofluorescence intensities in cell nuclei using digitised confocal image analysis were measured to semi-quantify differences in the pCREB immunoreactivity (pCREB-ir) caused by different treatments. Increasing concentrations of forskolin or 3-isobutyl-1-methylxanthine (IBMX) resulted in a dose-dependent rise of the mean fluorescence intensity in pCREB-ir nuclear staining. Concomitant melatonin application significantly decreased pCREB-ir in INS-1 cells after 30-min, 1-hr and 3-hr treatment. The melatonin receptor antagonists luzindole and 4-phenyl-2-propionamidotetraline (4P-PDOT) completely abolished the pCREB phosphorylation-decreasing effect of melatonin, indicating that both melatonin receptor isoforms (MT(1) and MT(2)) are involved. In a transfected INS-1 cell line expressing the human MT(2) receptor, melatonin caused the greatest reduction in pCREB after IBMX treatment compared with nontransfected INS-1 cells, indicating a crucial influence of melatonin receptor density on pCREB regulation. Furthermore, the downregulation of pCREB by melatonin is concomitantly associated with a statistically significant downregulation of Camk2d transcript levels, as measured after 3 hr. In conclusion, the present study provides evidence that the phosphorylation level of CREB is modulated in pancreatic β-cells by melatonin. Mediated via CREB, melatonin regulates the expression of genes that play an important functional role in the regulation of β-cell signalling pathways.
Sigma-Aldrich
Luzindole, ≥90%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00