MilliporeSigma
  • [Determination of 23 phthalate esters in food by solid-phase extraction coupled with gas chromatography-mass spectrometry].
[Determination of 23 phthalate esters in food by solid-phase extraction coupled with gas chromatography-mass spectrometry].
Se pu = Chinese journal of chromatography (2012-06-07)
Xianghua Zheng, Liyi Lin, Enhua Fang, Yonghui Huang, Shuang Zhou, Yu Zhou, Xiaoyan Zheng, Dunming Xu
ABSTRACT
A method for the simultaneous determination of 23 phthalate esters in food samples by solid-phase extraction coupled with gas chromatography-mass spectrometry (SPE-GC-MS) was developed and evaluated. The samples were extracted with hexane or acetonitrile, and cleaned up with a glass ProElut PSA SPE column. The identification and quantification were performed by GC-MS in selected ion monitoring (SIM) mode. The extraction processes of different foods were investigated. The calibration curves of phthalate esters showed good linearity in the range of 0.05-5 mg/L (0.5-5 mg/L for diisononyl phthalate (DINP), diisodecyl-phthalate (DIDP)) with the correlation coefficients (r) between 0.984 8 and 0.999 6. The limits of detection of phthalate esters in food samples ranged from 0.005 to 0.05 mg/kg (S/N = 3) and the limits of quantification ranged from 0.02 to 0.2 mg/kg (S/N = 10). The average recoveries of 23 analytes spiked in 10 kinds of food matrices ranged from 77% to 112% with the relative standard deviations (RSDs, n = 6) of 4.1%-12.5%. The method is suitable for the determination of 23 phthalate esters simultaneously in foodstuffs with easy operation, high accuracy and precision.
Sigma-Aldrich
Dimethyl phthalate, SAJ special grade, ≥99.0%
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Supelco
Dimethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Diethyl phthalate, SAJ special grade, ≥98.0%
Supelco
Diethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Diethyl phthalate, 99.5%
Sigma-Aldrich
Diethyl phthalate, ≥99%
Supelco
Diallyl phthalate, analytical standard
Sigma-Aldrich
Diallyl phthalate, 97%