MilliporeSigma
  • [A case of thyroid medullary carcinoma with multiple painful bone metastases successfully treated with strontium-89 chloride].
[A case of thyroid medullary carcinoma with multiple painful bone metastases successfully treated with strontium-89 chloride].
Gan to kagaku ryoho. Cancer & chemotherapy (2012-06-19)
Chiya Oshiro, Shunji Kamigaki, Takashi Arai, Yukio Nakamura, Mutsumi Fukunaga, Wakako Ichida, Hiroshi Ikeda
ABSTRACT
A 70-year-old man was diagnosed as thyroid medullary carcinoma with multiple bone metastases. He underwent total thyroidectomy and cervical lymph node dissection. After one year, the pain from his bone lesions was becoming severe. To relieve the pain, he was administered opioids and external-beam radiation therapy. However, he continued to have substantial multiple bone pain. We used combination therapy of strontium-89 chloride for the treatment of widespread multiple bone pain and external-beam radiation therapy for localized pain. That combination therapy was effective and improved the QOL of the patient. We used strontium-89 chloride four times within one year, and no serious side effects occurred during therapy. Our thoroughly investigated case suggests that strontium-89 therapy is one of the effective and safe therapies for patients with painful bone metastases of thyroid medullary carcinoma.
Sigma-Aldrich
Strontium chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Strontium chloride solution, 0.1 M
Sigma-Aldrich
Strontium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Strontium chloride hexahydrate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Strontium chloride hexahydrate, ACS reagent, 99%