MilliporeSigma
Partitioning behavior of petrodiesel/biodiesel blends in water.
Environmental science & technology (2012-06-22)
Mohamad H Yassine, Shuyun Wu, Makram T Suidan, Albert D Venosa
ABSTRACT
The partitioning behavior of six petrodiesel/soybean-biodiesel blends (B0, B20, B40, B60, B80, and B100, where B100 is 100% unblended biodiesel) in water was investigated at various oil loads by the 10-fold dilution method. Five fatty acid methyl esters (FAMEs), C10-C20 n-alkanes, and four monoaromatic compounds were targeted for analysis. Only the aromatic compounds were partitioned according to Raoult's law at all oil loads. The partitioning of the FAMEs and n-alkanes at higher oil loads was found to be orders of magnitude higher than the reported aqueous solubilities of these compounds, and directly correlated with the amount of oil load applied. Depth filtration of the water-accommodated fractions (WAFs) significantly reduced the observed concentrations of the FAMEs and n-alkanes, but did not appreciably affect the aromatic compounds. The FAMEs and n-alkanes concentrations in the filtered WAFs agreed with the aqueous solubilities of those compounds reported in the literature, but the n-alkanes showed progressive deviations from those solubilities with the increase in the amount of biodiesel in the blends. Further dilution experiments on pure n-hexadecane confirmed the presence of a metastable colloidal phase that seems to be controlled by hydrophobic interactions and surface phenomena. The addition of biodiesel to the oil blend appeared to have a positive impact on the dissolved concentrations and the colloidal accommodation of the n-alkanes. Autoxidation of the biodiesel constituents was found to be significant, and increased with increasing oil loads. Chemical products such as hexanal, n-butyl acetate, diethylene glycol monobutyl ether, and diethylene glycol monobutyl ether acetate were positively identified among the FAMEs' autoxidation byproducts. Our data suggest a positive enhancement for biodiesel on the formation of the oil in water colloidal phase, possibly by forming a surfactant-cosurfactant-like pair of the FAMEs and their autoxidation byproducts.
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Supelco
Hexadecane, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46