MilliporeSigma
  • Spectroscopic monitoring of photocatalytic degradation of the insecticide acetamiprid and its degradation product 6-chloronicotinic acid on TiO₂ catalyst.
Spectroscopic monitoring of photocatalytic degradation of the insecticide acetamiprid and its degradation product 6-chloronicotinic acid on TiO₂ catalyst.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2012-07-05)
Valéria Guzsvány, Ljiljana Rajić, Branislav Jović, Dejan Orčić, János Csanádi, Sanja Lazić, Biljana Abramović
ABSTRACT
Two spectroscopic methods, (1)H NMR and FTIR, were developed for the monitoring of the photocatalytic degradation of acetamiprid, a widely used pyridine-based neonicotinoid insecticide, in UV-irradiated aqueous suspensions of O(2)/TiO(2). The (1)H NMR method allowed also the identification of the intermediates such as 6-chloronicotinic and formic acids, as well as separate monitoring of the kinetics of degradation of acyclic and aromatic moieties based on the different chemical shifts of the protons belonging to the methyl group of the acyclic and selected proton of the heterocyclic aromatic moiety. The FTIR procedure enabled the monitoring of the kinetics of degradation of the cyano group of the compound. The obtained results are in good agreement with the comparative HPLC-DAD and HPLC-MS/MS measurements, which also enabled the identification of certain intermediates. To get a deeper insight into the complex photocatalytic process, the photocatalytic degradation of 6-chloronicotinic acid, a stable degradation intermediate of acetamiprid, was also investigated by (1)H NMR and HPLC-DAD methods. Based on the obtained data, a tentative reaction mechanism was proposed for the photocatalytic degradation of acetamiprid.
Sigma-Aldrich
6-Chloropyridine-3-carboxylic acid, 99%