MilliporeSigma
  • Innovations for combating dentin hypersensitivity: current state of the art.
Innovations for combating dentin hypersensitivity: current state of the art.
Compendium of continuing education in dentistry (Jamesburg, N.J. : 1995) (2012-07-11)
Yiming Li
ABSTRACT
Dentin hypersensitivity is a common dental complaint, and in severe cases it can impair eating, drinking, and even speaking, thus interfering with the normal daily life of an individual. Throughout years of research, there has been significant advancement in understanding dentin hypersensitivity, and various treatment regimens have been developed for combating the problem. The continued efforts have recently resulted in a novel technology that uses 8% arginine and calcium carbonate for treating dentin hypersensitivity. Formulations of oral care products using this technology--including toothpaste and prophylactic pastes--have demonstrated not only their effectiveness but also their ability to provide instant relief. This paper provides an overview of the etiology of dentin hypersensitivity, the discovery and development of measures for combating the problem, and the available data on the clinical efficacy of products based on 8% arginine and calcium carbonate.
Sigma-Aldrich
Potassium nitrate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Strontium chloride solution, 0.1 M
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Strontium chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Supelco
Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate, traceable to primary standards (LGC)
Sigma-Aldrich
Strontium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium fluorophosphate, 95%
Sigma-Aldrich
Strontium chloride hexahydrate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Strontium chloride hexahydrate, ACS reagent, 99%