MilliporeSigma
  • Inhibition of acetate accumulation leads to enhanced production of (R,R)-2,3-butanediol from glycerol in Escherichia coli.
Inhibition of acetate accumulation leads to enhanced production of (R,R)-2,3-butanediol from glycerol in Escherichia coli.
Journal of industrial microbiology & biotechnology (2012-07-27)
Xiaolin Shen, Yuheng Lin, Rachit Jain, Qipeng Yuan, Yajun Yan
ABSTRACT
This work describes the production of (R,R)-2,3-butanediol in Escherichia coli using glycerol by metabolic engineering approaches. The introduction of a synthetic pathway converting pyruvate to (R,R)-2,3-butanediol into wild-type E. coli strain BW25113 led to the production of (R,R)-2,3-butanediol at a titer of 3.54 g/l and a yield of 0.131 g product/g glycerol (26.7 % of theoretical maximum) with acetate (around 3.00 g/l) as the dominant by-product. We therefore evaluated the impacts of deleting the genes ackA or/and poxB that are responsible for the major by-product, acetate. This increased production of (R,R)-2,3-butanediol to 9.54 g/l with a yield of 0.333 g product/g glycerol (68.0 % of theoretical maximum) in shake flask studies. The utilization of low-priced crude glycerol to produce value-added chemicals is of great significance to the economic viability of the biodiesel industry.
Sigma-Aldrich
(2R,3R)-(−)-2,3-Butanediol, 97%
Sigma-Aldrich
(2S,3S)-(+)-2,3-Butanediol, 97%
Sigma-Aldrich
2,3-Butanediol, 98%
Sigma-Aldrich
meso-2,3-Butanediol, 99%