MilliporeSigma
  • Melatonin down-regulates volume-sensitive chloride channels in fibroblasts.
Melatonin down-regulates volume-sensitive chloride channels in fibroblasts.
Pflugers Archiv : European journal of physiology (2012-07-28)
Ismail Ben Soussia, Frédérique Mies, Robert Naeije, Vadim Shlyonsky
ABSTRACT
Melatonin has been reported to present with vasorelaxant and anti-fibrotic properties. We hypothesized that melatonin may down-regulate volume-regulated anion channels (VRAC) in fibroblasts to limit their migration and proliferation. While acute exposure of L929 fibroblasts to melatonin did not result in a significant decrease in VRAC current, pretreatment with 100 μM melatonin for 1 h decreased swelling-dependent activation of anion currents by 83% as measured by whole-cell perforated patch-clamp technique. This down-regulation of VRAC currents was dose-dependent with a half-maximal inhibition of 3.02 ± 0.48 μM. Overnight treatment of cells with 100 nM melatonin had the same inhibitory potency as a 1-h treatment with 100 μM. A similar down-regulatory effect of melatonin on VRAC was observed in primary rat lung fibroblasts. The effect of melatonin was prevented by luzindole and K185 that suggests implication of MT2 receptor. GF109203X, a protein kinase C inhibitor, blocked melatonin's action on VRAC, indicating that MT2 receptor activation results in stimulation of PKC. Consequently, melatonin inhibited regulatory volume decrease following hypotonic swelling of cells. Melatonin also decreased the migration of L929 fibroblasts through the same pathways that blocked VRAC. There was no significant inhibition of cell proliferation. Our study suggests that the attenuation of fibrosis and vascular remodeling by melatonin seen in animal models of hypertension and pulmonary fibrosis might be, in part, related to blunted fibroblast migration possibly through protein kinase C-mediated decrease in chloride channel activity.
Sigma-Aldrich
GF 109203X, synthetic, ≥90% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Luzindole, ≥90%