MilliporeSigma
  • Metal-free, NHPI catalyzed oxidative cleavage of C-C double bond using molecular oxygen as oxidant.
Metal-free, NHPI catalyzed oxidative cleavage of C-C double bond using molecular oxygen as oxidant.
Organic letters (2012-08-03)
Riyuan Lin, Feng Chen, Ning Jiao
ABSTRACT
A metal-free N-hydroxyphthalimide (NHPI) catalyzed aerobic oxidative cleavage of olefins has been developed. Molecular oxygen is used as the oxidant and reagent for this oxygenation reaction. This methodology has prevented the use of toxic metals or overstoichiometric amounts of traditional oxidants, showing good economical and environmental advantages. Based on the experimental observations, a plausible mechanism is proposed.
Sigma-Aldrich
N-Hydroxyphthalimide, 97%