MilliporeSigma
  • 1D coaxial platinum/titanium nitride nanotube arrays with enhanced electrocatalytic activity for the oxygen reduction reaction: towards Li-air batteries.
1D coaxial platinum/titanium nitride nanotube arrays with enhanced electrocatalytic activity for the oxygen reduction reaction: towards Li-air batteries.
ChemSusChem (2012-08-07)
Shanmu Dong, Xiao Chen, Shan Wang, Lin Gu, Lixue Zhang, Xiaogang Wang, Xinhong Zhou, Zhihong Liu, Pengxian Han, Yulong Duan, Hongxia Xu, Jianhua Yao, Chuanjian Zhang, Kejun Zhang, Guanglei Cui, Liquan Chen
ABSTRACT
CAT ON A HOT TIN SUPPORT: Coaxial Pt/TiN nanotube arrays are used to achieve a superior electrocatalytic activity of platinum towards the oxygen reduction reaction (ORR). Compared to a commercial Pt/C catalyst, the Pt/TiN NTA materials delivers a higher mass activity and specific activity for the ORR. Hence, these materials are useful as cathodes for hybrid electrolyte Li-air batteries, as demonstrated.
Sigma-Aldrich
Titanium nitride, <3 μm