Merck
  • A new member of the oxygen-photosensitizers family: a water-soluble polymer binding a platinum complex.
A new member of the oxygen-photosensitizers family: a water-soluble polymer binding a platinum complex.
Dalton transactions (Cambridge, England : 2003) (2012-08-14)
Loredana Ricciardi, Francesco Puoci, Giuseppe Cirillo, Massimo La Deda
ABSTRACT
The grafting of a 2-picolylamine Pt(II) complex into polymethacrylic acid has been successfully performed. The obtained polymer is water soluble, and it represents the first example of a platinum-containing polymer able to photogenerate singlet oxygen.
Sigma-Aldrich
Poly(methacrylic acid, sodium salt) solution, average Mn ~5,400, average Mw ~9,500 by GPC, 30 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(methacrylic acid, sodium salt) solution, average Mw 4,000-6,000, 40 wt. % in H2O