MilliporeSigma
  • Threshold voltage fluctuation in 16-nm-gate FinFets induced by random work function of nanosized metal grain.
Threshold voltage fluctuation in 16-nm-gate FinFets induced by random work function of nanosized metal grain.
Journal of nanoscience and nanotechnology (2012-08-22)
Yiming Li, Hui-Wen Cheng, Chi-Hong Hwang
ABSTRACT
The random work-function (WK) induced threshold voltage fluctuation (sigmaVth) in 16 nm Titanium Nitride (TiN) metal-gate fin-type field effect transistors (FinFETs) is explored and modeled by using an experimentally validated Monte Carlo simulation approach. The influences of metal-grain size and device geometry aspect ratio on the random WK-induced sigmaVth are considered in the proposed equation analytically. The formula accounts for the inside of fluctuation and could be used for the assessment of effectiveness of suppression techniques.
Sigma-Aldrich
Titanium nitride, <3 μm