MilliporeSigma
  • Current-confinement structure and extremely high current density in organic light-emitting transistors.
Current-confinement structure and extremely high current density in organic light-emitting transistors.
Advanced materials (Deerfield Beach, Fla.) (2012-09-11)
Kosuke Sawabe, Masaki Imakawa, Masaki Nakano, Takeshi Yamao, Shu Hotta, Yoshihiro Iwasa, Taishi Takenobu
ABSTRACT
Extremely high current densities are realized in single-crystal ambipolar light-emitting transistors using an electron-injection buffer layer and a current-confinement structure via laser etching. Moreover, a linear increase in the luminance was observed at current densities of up to 1 kA cm(-2) , which is an efficiency-preservation improvement of three orders of magnitude over conventional organic light-emitting diodes (OLEDs) at high current densities.
Sigma-Aldrich
2,2′:5′,2′′-Terthiophene, 99%