MilliporeSigma
  • A new entry to the construction of a quaternary carbon center having a fluorine atom--S(N)2' reaction of γ-fluoroallylic alcohol derivatives with various cyanocuprates.
A new entry to the construction of a quaternary carbon center having a fluorine atom--S(N)2' reaction of γ-fluoroallylic alcohol derivatives with various cyanocuprates.
Organic & biomolecular chemistry (2012-09-12)
Tsutomu Konno, Atsushi Ikemoto, Takashi Ishihara
ABSTRACT
Treatment of γ-fluoroallylic phosphate with various lower-ordered cyanocuprates derived from Grignard reagents, organolithium, and organozincs gave the corresponding S(N)2' products having a fluorine atom at a quaternary carbon center in excellent yields. This system could be successfully extended to the chiral version, enantiomerically pure fluorine-containing materials also being obtained in high yield.
Sigma-Aldrich
Allyl alcohol, ≥98.5%
Sigma-Aldrich
Allyl alcohol, ≥98.5%
Sigma-Aldrich
Allyl alcohol, ≥99%