MilliporeSigma
Antioxidant properties of Mannich bases.
Bioorganic & medicinal chemistry letters (2012-09-22)
Dong Ho Park, Jayachandran Venkatesan, Se-Kwon Kim, Venkatachalm Ramkumar, Paramasivam Parthiban
ABSTRACT
The biological importance of antioxidants influenced to synthesize some curcumin-related compounds as potential antioxidants. Accordingly, a series of 2,4-diaryl-3-azabicyco[3.3.1]nonan-9-ones were synthesized with polyphenolic and/or polymethoxyphenyl groups by modified Mannich condensations. The yield was significantly improved using BF(3)·SiO(2) as heterogeneous catalyst under mild conditions. Stereochemistry of all the synthesized compounds was established as twin-chair with an equatorial disposition of the aryl groups, through their NMR and XRD interpretations. The ABNs 8 (curcumin analog) and 10 (bis-demethoxycurcumin analog) showed an effective profile over curcumin, α-tocopherol, and vitamin C by chemical methods. Further, the efficiency of one of the active molecules, ABN 10, was demonstrated by its intracellular ROS inhibition activity on RAW 264.7 macrophage cells by FACS analysis in dose-dependent manner.
Sigma-Aldrich
Boron trifluoride, electronic grade, ≥99.99%
Supelco
Boron trifluoride - 1-butanol solution, ~10% in 1-butanol (∼1.3 M), for GC derivatization, LiChropur