MilliporeSigma
  • Rapid and simultaneous determination of twenty amino acids in complex biological and food samples by solid-phase microextraction and gas chromatography-mass spectrometry with the aid of experimental design after ethyl chloroformate derivatization.
Rapid and simultaneous determination of twenty amino acids in complex biological and food samples by solid-phase microextraction and gas chromatography-mass spectrometry with the aid of experimental design after ethyl chloroformate derivatization.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-09-25)
Mohana Krishna Reddy Mudiam, Ch Ratnasekhar, Rajeev Jain, Prem Narain Saxena, Abhishek Chauhan, R C Murthy
ABSTRACT
Amino acids play a vital role as intermediates in many important metabolic pathways such as the biosynthesis of nucleotides, vitamins and secondary metabolites. A sensitive and rapid analytical method has been proposed for the first time for the simultaneous determination of twenty amino acids using solid-phase microextraction (SPME). The protein samples were hydrolyzed by 6M HCl under microwave radiation for 120 min. Then the amino acids were derivatized by ethyl chloroformate (ECF) and the ethoxy carbonyl ethyl esters of amino acids formed were extracted using SPME by direct immersion. Finally the extracted analytes on the SPME fiber were desorbed at 260°C and analyzed by gas chromatography-mass spectrometer (GC-MS) in electron ionization mode. Factors which affect the SPME efficiency were screened by Plackett-Burmann design; most significant factors were optimized with response surface methodology. The optimum conditions for SPME are as follows: pH of 1.7, ionic strength of 733 mg, extraction time of 30 min and fiber of divinyl benzene/carboxen/polydimethylsiloxane (DVB/CAR/PDMS). The recovery of all the amino acids was found to be in the range of 89.17-100.98%. The limit of detection (LOD) of all derivatized amino acids in urine, hair and soybean was found to be in the range of 0.20-7.52 μg L(-1), 0.21-8.40 μg L(-1) and 0.18-5.62 μg L(-1), respectively. Finally, the proposed technique was successfully applied for the determination of amino acids in complex biological (hair, urine) and food samples (soybean). The method can find wide applications in the routine analysis of amino acids in any biological as well as food samples.
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Supelco
ORBO-93 Sorbent Tube, ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium hydrogen sulfate, technical grade
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Sigma-Aldrich
Sodium sulfate, JIS special grade, ≥99.0%
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Supelco
SPME Fiber Assembly, matrix active group 50/30µm DVB/CAR/PDMS, StableFlex fiber, manual holder, needle size 24 ga
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME Fiber Assembly, for use with autosampler, matrix active group 50/30µm DVB/CAR/PDMS, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler