MilliporeSigma
  • Enzyme-linked immunoassay. Conjugation of rabbit anti-(human immunoglobulin G) antibody with beta-D-galactosidase from Escherichia coli and its use for human immunoglobulin G assay.
Enzyme-linked immunoassay. Conjugation of rabbit anti-(human immunoglobulin G) antibody with beta-D-galactosidase from Escherichia coli and its use for human immunoglobulin G assay.
European journal of biochemistry (1976-02-16)
K Kato, Y Hamaguchi, H Fukui, E Ishikawa
PMID943288
ABSTRACT
Rabbit immunoglobulin G (IgG) was reduced by incubating with 10 mM 2-mercaptoethylamine and then treated with excess amounts of N,N'-o-phenylenedimaleimide. As a result, maleimide residues were introduced into rabbit IgG molecules. Rabbit IgG containing maleimide residues could be coupled to beta-D-galactosidase from Escherichia coli which has sulfhydryl groups in the molecule. The resulting rabbit IgG (antibody)-enzyme complex may be useful for immunoassay of antigens. As an example, human IgG was assayed by the sandwich method using the rabbit anti-(human IgG) IgG-beta-D-galactosidase complex and amounts of human IgG as small as 3 fmoles were measurable.
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 kg
Estimated to ship on March 06, 2025
$2,860.00