MilliporeSigma
Enzyme immunoassay of thyrotropin releasing hormone (TRH).
Chemical & pharmaceutical bulletin (1992-08-01)
M Takeyama, A Otaka, N Fujii
ABSTRACT
A sensitive and specific double-antibody enzyme immunoassay (EIA) for a thyrotropin releasing hormone (TRH)-like immunoreactive substance has been developed. In order to synthesize TRH-labeled beta-D-galactosidase (beta-gal), a newly devised TRH derivative, pGlu-His-Pro-NH-(CH2)6-NH2 (TRH-Hex), was employed. TRH-Hex was linked to beta-gal by the N-(epsilon-maleimidocaproyloxy) succinimide coupling procedure. For competitive reactions, the TRH antibody was incubated with standard TRH and TRH-Hex-beta-gal (delayed addition). Free and antibody-bound enzyme hapten were separated by using an anti-rabbit immunoglobulin G coated immunoplate. Activity of the enzyme on the plate was fluorometrically determined. The present immunoassay allows detection of 0.8 to 100 pmol/well of TRH.
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
In Stock
Details...
₩150,220
500 g
Please contact Customer Service for Availability
₩448,427